Zirconium(IV) oxide 99+%
Supplier: Apollo Scientific
Warning
Synonyms: Zirconium dioxide, Zirconium Oxide
IN3922-1KG IN3922-250G
APOSIN3922-1KGEA 0 EUR
APOSIN3922-1KG APOSIN3922-250G
Zirconium(IV) oxide 99+%
Zirconium (IV) oxide
Formula: ZrO₂
MW: 123.22 g/mol
Boiling Pt: 4300
Melting Pt: 2715 °C
Density: 5.89 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011310
CAS Number: 1314-23-4
Merck Index: 14,10180