5-Amino-2-methylphenol
Supplier: Apollo Scientific
Warning
Synonyms: 5-Amino-o-cresol, 3-Hydroxy-4-methylaniline, 1-Amino-3-hydroxy-4-methylbenzene, 5-Amino-2-methylphenol, 4-Amino-2-hydroxytoluene
OR0011-100G OR0011-25G
APOSOR0011-100GEA 26.1 EUR
APOSOR0011-100G APOSOR0011-25G
5-Amino-2-methylphenol
5-Amino-2-methylphenol
Formula: C₇H₉NO
Melting Pt: 160…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00043922
CAS Number: 2835-95-2
EINECS: 220-618-6