1-Bromo-2-chlorobenzene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromochlorobenzene
OR0017-100G OR0017-250G OR0017-25G
APOSOR0017-100GEA 45.1 EUR
APOSOR0017-100G APOSOR0017-250G APOSOR0017-25G
1-Bromo-2-chlorobenzene 99%
1-Bromo-2-chlorobenzene
Formula: C₆H₄BrCl
Boiling Pt: 203…205 °C
Melting Pt: –13 °C
Density: 1.649 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00000532
CAS Number: 694-80-4
EINECS: 211-775-1