3-Chlorostyrene 97%

Supplier: Apollo Scientific
Warning

OR0030-25G OR0030-5G
APOSOR0030-25GEA 113 EUR
APOSOR0030-25G APOSOR0030-5G
3-Chlorostyrene 97%
3-Chlorostyrene
Formula: C₈H₇Cl
Boiling Pt: 188…189 °C
Density: 1.09 g/cm³ (25 °C)
Flash Pt: 67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000598
CAS Number: 2039-85-2
EINECS: 218-024-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR