4-Nitro-o-anisidine 98%
Supplier: Apollo Scientific
|
Warning
|
OR0045-1KG
OR0045-500G
APOSOR0045-1KGEA
94.4
EUR
APOSOR0045-1KG
APOSOR0045-500G
4-Nitro-o-anisidine 98%
4-Nitro-o-anisidine
Formula:
C₇H₈N₂O₃ Melting Pt: 138…142 °C Density: 1.211 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007363 CAS Number: 97-52-9 EINECS: 202-588-6 |