3-Amino-2,6-dichloropyridine 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2,6-Dichloro-3-pyridinamine, 2,6-Dichloropyridin-3-amine, 2,6-Dichloro-3-pyridylamine
OR0126-100G OR0126-25G OR0126-5G
APOSOR0126-100GEA 188 EUR
APOSOR0126-100G APOSOR0126-25G APOSOR0126-5G
3-Amino-2,6-dichloropyridine 98%
3-Amino-2,6-dichloropyridine
Formula: C₅H₄Cl₂N₂
Melting Pt: 120…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00023417
CAS Number: 62476-56-6
EINECS: 263-559-1
UN: 2811
ADR: 6.1,III