2-Oxa-5-azabicyclo[2.2.1]heptane hydrochloride
Supplier: Apollo Scientific
Warning
OR015021-250MG OR015021-1G
APOSOR015021-250MGEA 26.1 EUR
APOSOR015021-250MG APOSOR015021-1G
2-Oxa-5-azabicyclo[2.2.1]heptane hydrochloride
2-Oxa-5-azabicyclo[2.2.1]heptane hydrochloride
Formula: C₅H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD07798642
CAS Number: 909186-56-7