3-Ethoxyacryloyl chloride 90%
Supplier: Apollo Scientific
Danger
OR015045-5G OR015045-1G OR015045-25G
APOSOR015045-5GEA 227 EUR
APOSOR015045-5G APOSOR015045-1G APOSOR015045-25G
3-Ethoxyacryloyl chloride 90%
3-Ethoxyacryloyl chloride
Formula: C₅H₇ClO₂
Boiling Pt: 75 °C
Density: 1.14 g/cm³ (25 °C)
Flash Pt: 25 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09029675
CAS Number: 6191-99-7
EINECS: 228-239-8
UN: 2920
ADR: 8+3,II