2-Phenoxy-N-methylethylamine hydrochloride 95+%
Supplier: Apollo Scientific
OR0179-1G OR0179-5G OR0179-25G
APOSOR0179-1GEA 185 EUR
APOSOR0179-1G APOSOR0179-5G APOSOR0179-25G
2-Phenoxy-N-methylethylamine hydrochloride 95+%
2-Phenoxy-N-methylethylamine hydrochloride
Formula: C₉H₁₄ClNO
Melting Pt: 174…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00460774
CAS Number: 85262-76-6