2-methylpropylhydrazine hydrochloride 97%
Supplier: Apollo Scientific
Warning
Synonyms: Isobutylhydrazine hydrochloride
OR0203-5G OR0203-25G
APOSOR0203-5GEA 166 EUR
APOSOR0203-5G APOSOR0203-25G
2-methylpropylhydrazine hydrochloride 97%
2-methylpropylhydrazine hydrochloride
Formula: C₄H₁₃ClN₂
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD06245510
CAS Number: 237064-47-0
UN: 2811
ADR: 6.1,III