4-Ethynyltoluene
Supplier: Apollo Scientific
Warning
OR0642-25G OR0642-5G OR0642-100G
APOSOR0642-25GEA 92.8 EUR
APOSOR0642-25G APOSOR0642-5G APOSOR0642-100G
4-Ethynyltoluene
4-Ethynyltoluene
Formula: C₉H₈
Boiling Pt: 168…170 °C
Melting Pt: 23 °C
Density: 0.916 g/cm³ (25 °C)
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00008571
CAS Number: 766-97-2
UN: 3295
ADR: 3,III