2-(Methylthio)benzylamine
Supplier: Apollo Scientific
Danger
OR0657-1G OR0657-250MG
APOSOR0657-1GEA 232 EUR
APOSOR0657-1G APOSOR0657-250MG
2-(Methylthio)benzylamine
2-(Methylthio)benzylamine
Formula: C₈H₁₁NS
Boiling Pt: 139…141 °C/15mmHg
Storage Temperature: Ambient
MDL Number: MFCD00831039
CAS Number: 56004-83-2
UN: 1760
ADR: 8,III