(2S)-(+)-2,3-Epoxypropyl toluene-4-sulphonate 98%
Supplier: Apollo Scientific
OR0739-100G OR0739-25G
APOSOR0739-100GEA 142 EUR
APOSOR0739-100G APOSOR0739-25G
(2S)-(+)-2,3-Epoxypropyl toluene-4-sulphonate 98%
(2S)-(+)-2,3-Epoxypropyl toluene-4-sulphonate
Formula: C₁₀H₁₂O₄S
Melting Pt: 47…49 °C
Storage Temperature: Ambient
MDL Number: MFCD00064489
CAS Number: 70987-78-9