4-(Boc-amino)piperidine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-N-(tert-Butoxycarbonyl)aminopiperidine, tert-Butyl piperidin-4-ylcarbamate, tert-butyl N-(piperidin-4-yl)carbamate, 4-Boc-Aminopiperidine, 4-N-BOC-Aminopiperidine
OR0807-25G OR0807-100G
APOSOR0807-25GEA 37.7 EUR
APOSOR0807-25G APOSOR0807-100G
4-(Boc-amino)piperidine 97%
4-(Boc-amino)piperidine
Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Melting Pt: 162…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00798171
CAS Number: 73874-95-0