2-Bromo-5-methylpyridine
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-5-picoline
OR0845-100G OR0845-25G
APOSOR0845-100GEA 43.5 EUR
APOSOR0845-100G APOSOR0845-25G
2-Bromo-5-methylpyridine
2-Bromo-5-methylpyridine
Formula: C₆H₆BrN
Boiling Pt: 95…96 °C/12.5mmHg
Melting Pt: 40…45 °C
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00209553
CAS Number: 3510-66-5