3-Bromobenzylamine hydrochloride 98%
Supplier: Apollo Scientific
Warning
Synonyms: (3-Bromophenyl)methanamine hydrochloride
OR0852-25G OR0852-5G OR0852-100G
APOSOR0852-25GEA 84.1 EUR
APOSOR0852-25G APOSOR0852-5G APOSOR0852-100G
3-Bromobenzylamine hydrochloride 98%
3-Bromobenzylamine hydrochloride
Formula: C₇H₉BrClN
MW: 222.52 g/mol
Melting Pt: 218…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00012856
CAS Number: 39959-54-1