4-(Dimethylamino)piperidine
Supplier: Apollo Scientific
Danger
Synonyms: N,N-Dimethylpiperidin-4-amine
OR0876-1G OR0876-25G OR0876-5G
APOSOR0876-1GEA 21.9 EUR
APOSOR0876-1G APOSOR0876-25G APOSOR0876-5G
4-(Dimethylamino)piperidine
4-(Dimethylamino)piperidine
Formula: C₇H₁₆N₂
Boiling Pt: 187 °C
Density: 1.09 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00023144
CAS Number: 50533-97-6
EINECS: 256-617-2
UN: 2735
ADR: 8,III