Benzopinacol

Supplier: Apollo Scientific
Warning

Synonyms: 1,1,2,2-Tetraphenyl-1,2-ethanediol, 1,1,2,2-Tetraphenylethane-1,2-diol, Tetraphenylethylene glycol, Tetraphenyl-1,2-ethanediol

OR0902-100G OR0902-25G OR0902-500G
APOSOR0902-100GEA 43.5 EUR
APOSOR0902-100G APOSOR0902-25G APOSOR0902-500G
Benzopinacol
Benzopinacol
Formula: (C₆H₅)₂C(OH)C(OH)(C₆H₅)₂
Melting Pt: 171…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00004448
CAS Number: 464-72-2
EINECS: 207-356-8
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR