2-Amino-6-chlorobenzothiazole
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Chlorobenzothiazol-2-ylamine, 6-Chloro-1,3-benzothiazol-2-amine
OR0921-100G
OR0921-25G
APOSOR0921-100GEA
37.7
EUR
APOSOR0921-100G
APOSOR0921-25G
2-Amino-6-chlorobenzothiazole
2-Amino-6-chlorobenzothiazole
Formula:
C₇H₅ClN₂S Melting Pt: 199…201 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053557 CAS Number: 95-24-9 EINECS: 202-402-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...