Zinc-copper couple
Supplier: Apollo Scientific
Danger
OR100-100G OR100-250G OR100-1KG
APOSOR100-100GEA 120 EUR
APOSOR100-100G APOSOR100-250G APOSOR100-1KG
Zinc-copper couple
Zinc-copper couple
Formula: CuZn
MW: 128.926 g/mol
Melting Pt: 419…420 °C
Storage Temperature: Ambient
MDL Number: MFCD00084851
CAS Number: 53801-63-1
UN: 3089
ADR: 4.1 III