(3-Bromopropyl)benzene 95%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-3-phenylpropane
OR10084-25G OR10084-1G OR10084-5G
APOSOR10084-25GEA 29 EUR
APOSOR10084-25G APOSOR10084-1G APOSOR10084-5G
(3-Bromopropyl)benzene 95%
(3-Bromopropyl)benzene
Formula: BrH₂CCH₂CH₂(C₆H₅)
MW: 199.09 g/mol
Boiling Pt: 219…220 °C
Density: 1.31 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000257
CAS Number: 637-59-2
EINECS: 211-294-7