Chlorocyclopentane
Supplier: Apollo Scientific
Danger
OR10099-100G OR10099-500G
APOSOR10099-100GEA 0 EUR
APOSOR10099-100G APOSOR10099-500G
Chlorocyclopentane
Chlorocyclopentane
Formula: C₅H₉Cl
Boiling Pt: 113…114 °C
Melting Pt: –94 °C
Density: 1.005 g/cm³ (25 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00001360
CAS Number: 930-28-9
EINECS: 213-212-5
UN: 1993
ADR: 3,II