2,4,6-Trichlorobenzenesulphonamide
Supplier: Apollo Scientific
Warning
OR1014-1G OR1014-5G
APOSOR1014-1GEA 43.5 EUR
APOSOR1014-1G APOSOR1014-5G
2,4,6-Trichlorobenzenesulphonamide
2,4,6-Trichlorobenzenesulphonamide
Formula: C₆H₄Cl₃NO₂S
Melting Pt: 179…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00955746
CAS Number: 28460-30-2