2-(1H-Pyrazol-4-yl)pyrazine
Supplier: Apollo Scientific
Warning
OR1028-1G OR1028-5G
APOSOR1028-1GEA 161 EUR
APOSOR1028-1G APOSOR1028-5G
2-(1H-Pyrazol-4-yl)pyrazine
2-(1H-Pyrazol-4-yl)pyrazine
Formula: C₇H₆N₄
MW: 146.15 g/mol
Melting Pt: 171…174 °C
Storage Temperature: Ambient
MDL Number: MFCD02579320
CAS Number: 849924-97-6