2,4-Dichloro-5-thiazolecarboxaldehyde 95%
Supplier: Apollo Scientific
Danger
Synonyms: 2,4-Dichlorothiazole-5-carbaldehyde
OR103038-1G OR103038-5G OR103038-25G
APOSOR103038-1GEA 108 EUR
APOSOR103038-1G APOSOR103038-5G APOSOR103038-25G
2,4-Dichloro-5-thiazolecarboxaldehyde 95%
2,4-Dichloro-5-thiazolecarboxaldehyde
Formula: C₄HCl₂NOS
MW: 182.03 g/mol
Boiling Pt: 180 °C/20mmHg
Melting Pt: 47…53 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00793013
CAS Number: 92972-48-0
UN: 2811
ADR: 6.1 III