2-(1H-Pyrazol-4-yl)quinoline 95+%
Supplier: Apollo Scientific
Warning
OR1033-1G OR1033-5G OR1033-10G
APOSOR1033-1GEA 148 EUR
APOSOR1033-1G APOSOR1033-5G APOSOR1033-10G
2-(1H-Pyrazol-4-yl)quinoline 95+%
2-(1H-Pyrazol-4-yl)quinoline
Formula: C₁₂H₉N₃
Melting Pt: 182…183 °C
Storage Temperature: Ambient
MDL Number: MFCD02579322
CAS Number: 439106-87-3