4-Aminotetrahydropyran hydrochloride 97%
Supplier: Apollo Scientific
Warning
Synonyms: Tetrahydro-2H-pyran-4-amine hydrochloride
OR10343-1G OR10343-25G
APOSOR10343-1GEA 21.9 EUR
APOSOR10343-1G APOSOR10343-25G
4-Aminotetrahydropyran hydrochloride 97%
4-Aminotetrahydropyran hydrochloride
Formula: C₅H₁₂ClNO
MW: 137.6079 g/mol
Melting Pt: 230…250 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00100881
CAS Number: 33024-60-1