1,4-Butanesultam
Supplier: Apollo Scientific
Warning
OR10390-1G OR10390-250MG
APOSOR10390-1GEA 81.2 EUR
APOSOR10390-1G APOSOR10390-250MG
1,4-Butanesultam
1,4-Butanesultam
Formula: C₄H₉NO₂S
Melting Pt: 94…96 °C
Storage Temperature: Ambient
MDL Number: MFCD04038916
CAS Number: 37441-50-2