Indole-4-carbaldehyd 98%
Supplier: Apollo Scientific
Warning
OR10487-100G OR10487-25G OR10487-5G
APOSOR10487-100GEA 538 EUR
APOSOR10487-100G APOSOR10487-25G APOSOR10487-5G
Indole-4-carbaldehyd 98%
Indole-4-carbaldehyd
Formula: C₉H₇NO
Melting Pt: 139…145 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01632221
CAS Number: 1074-86-8