Cloxiquina 98%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Chloro-8-quinolinol, 5-Chloroquinolin-8-ol, 5-Chloro-8-hydroxyquinoline
OR1049-100G OR1049-500G
APOSOR1049-100GEA 24.8 EUR
APOSOR1049-100G APOSOR1049-500G
Cloxiquina 98%
Cloxiquina
Formula: C₉H₆ClNO
Melting Pt: 121…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00006788
CAS Number: 130-16-5
EINECS: 204-978-1