2,4,5-Triiodoimidazole
Supplier: Apollo Scientific
Warning
Synonyms: 2,4,5-Triiodo-1H-imidazole
OR10591-5G OR10591-1G OR10591-10G
APOSOR10591-5GEA 31.9 EUR
APOSOR10591-5G APOSOR10591-1G APOSOR10591-10G
2,4,5-Triiodoimidazole
2,4,5-Triiodoimidazole
Formula: C₃HI₃N₂
Melting Pt: 186…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00068812
CAS Number: 1746-25-4