1,3-Dibromo-2-propanol
Supplier: Apollo Scientific
Danger
Synonyms: Glycerol alpha,alpha'-Dibromohydrin, 1,3-Dibromopropan-2-ol, alpha,alpha'-Dibromohydrin
OR1061-1G OR1061-25G OR1061-5G
APOSOR1061-1GEA 29 EUR
APOSOR1061-1G APOSOR1061-25G APOSOR1061-5G
1,3-Dibromo-2-propanol
1,3-Dibromo-2-propanol
Formula: BrH₂CCH(OH)CH₂Br
Boiling Pt: 215 °C
Density: 2.136 g/cm³ (25 °C)
Flash Pt: 47 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000216
CAS Number: 96-21-9
EINECS: 202-489-8
UN: 1992
ADR: 3+6.1,III