(Methoxycarbonylmethyl)triphenylphosphonium chloride
Supplier: Apollo Scientific
Warning
OR10662-50G OR10662-1KG OR10662-250G
APOSOR10662-50GEA 21.9 EUR
APOSOR10662-50G APOSOR10662-1KG APOSOR10662-250G
(Methoxycarbonylmethyl)triphenylphosphonium chloride
(Methoxycarbonylmethyl)triphenylphosphonium chloride
Formula: C₂₁H₂₀ClO₂P
MW: 370.82 g/mol
Melting Pt: 155 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00050199
CAS Number: 2181-97-7