3-Chloroanisole
Supplier: Apollo Scientific
Warning
OR1078-250G OR1078-50G OR1078-1KG
APOSOR1078-250GEA 106 EUR
APOSOR1078-250G APOSOR1078-50G APOSOR1078-1KG
3-Chloroanisole
3-Chloroanisole
Formula: C₇H₇ClO
Boiling Pt: 193 °C
Density: 1.164 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00000591
CAS Number: 2845-89-8
EINECS: 220-642-7