4-Hydroxybenzonitrile 98%
Supplier: Apollo Scientific
Warning
Synonyms: p-Cyanophenol, 4-Cyanophenol, p-Hydroxybenzonitrile
OR10907-100G OR10907-500G OR10907-25G
APOSOR10907-100GEA 40.6 EUR
APOSOR10907-100G APOSOR10907-500G APOSOR10907-25G
4-Hydroxybenzonitrile 98%
4-Hydroxybenzonitrile
Formula: C₇H₅NO
MW: 119.12 g/mol
Boiling Pt: 146 °C/2mmHg
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002312
CAS Number: 767-00-0
EINECS: 212-175-2