3-Mercaptophenylboronic acid
Supplier: Apollo Scientific
Warning
OR10936-1G OR10936-5G
APOSOR10936-1GEA 144 EUR
APOSOR10936-1G APOSOR10936-5G
3-Mercaptophenylboronic acid
3-Mercaptophenylboronic acid
Formula: C₆H₇BO₂S
Melting Pt: 196…201 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095131
CAS Number: 352526-01-3