2-Furoic acid 95%
Supplier: Apollo Scientific
Warning
Synonyms: Furan-2-carboxylic acid, 2-Furancarboxylic acid
OR10949-100G OR10949-25G OR10949-500G
APOSOR10949-100GEA 24.8 EUR
APOSOR10949-100G APOSOR10949-25G APOSOR10949-500G
2-Furoic acid 95%
2-Furoic acid
Formula: C₅H₄O₃
Boiling Pt: 230 °C
Melting Pt: 127…132 °C
Flash Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD00003238
CAS Number: 88-14-2
EINECS: 201-803-0