2-Amino-4-(4-cyanophenyl)thiazole
Supplier: Apollo Scientific
Warning
Synonyms: 4-(2-Amino-1,3-thiazol-4-yl)benzonitrile
OR110560-1G OR110560-5G OR110560-10G
APOSOR110560-1GEA 137 EUR
APOSOR110560-1G APOSOR110560-5G APOSOR110560-10G
2-Amino-4-(4-cyanophenyl)thiazole
2-Amino-4-(4-cyanophenyl)thiazole
Formula: C₁₀H₇N₃S
Melting Pt: 257…268 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03407828
CAS Number: 436151-85-8