2-Ethyl-1,3-benzoxazol-6-amine
Supplier: Apollo Scientific
Warning
OR110721-1G OR110721-5G
APOSOR110721-1GEA 116 EUR
APOSOR110721-1G APOSOR110721-5G
2-Ethyl-1,3-benzoxazol-6-amine
2-Ethyl-1,3-benzoxazol-6-amine
Formula: C₉H₁₀N₂O
Melting Pt: 83…85 °C
Storage Temperature: Ambient
CAS Number: 855423-35-7