Methyl-5-nitro-2-(1-pyrrolidinyl)benzenecarboxylate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 5-nitro-2-pyrrolidin-1-ylbenzoate
OR110838-1G OR110838-5G OR110838-10G
APOSOR110838-1GEA 21.9 EUR
APOSOR110838-1G APOSOR110838-5G APOSOR110838-10G
Methyl-5-nitro-2-(1-pyrrolidinyl)benzenecarboxylate
Methyl-5-nitro-2-(1-pyrrolidinyl)benzenecarboxylate
Formula: C₁₂H₁₄N₂O₄
MW: 250.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00662491
CAS Number: 259269-68-6