4-Bromo-N,N-dimethylaniline
Supplier: Apollo Scientific
Warning
OR11136-100G OR11136-500G
APOSOR11136-100GEA 0 EUR
APOSOR11136-100G APOSOR11136-500G
4-Bromo-N,N-dimethylaniline
4-Bromo-N,N-dimethylaniline
Formula: C₈H₁₀BrN
Boiling Pt: 264 °C
Melting Pt: 52…54 °C
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000093
CAS Number: 586-77-6
EINECS: 209-582-2