4-Methylindole 96%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Methyl-1H-indole
OR1114-1G OR1114-25G OR1114-5G
APOSOR1114-1GEA 21.9 EUR
APOSOR1114-1G APOSOR1114-25G APOSOR1114-5G
4-Methylindole 96%
4-Methylindole
Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 267 °C
Melting Pt: 5 °C
Density: 1.06 g/cm³ (25 °C)
Flash Pt: >112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005668
CAS Number: 16096-32-5
EINECS: 240-262-5