2-bromo-1-(o-tolyl)ethanone
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-1-(o-tolyl)ethan-1-one, 2-Bromo-2'-methylacetophenone
OR11269-1G OR11269-5G OR11269-25G
APOSOR11269-1GEA 36.4 EUR
APOSOR11269-1G APOSOR11269-5G APOSOR11269-25G
2-bromo-1-(o-tolyl)ethanone
2-Bromo-1-(o-tolyl)ethanone
Formula: C₉H₉BrO
MW: 213.074 g/mol
Boiling Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD04038955
CAS Number: 51012-65-8