(6-Chloro-3-pyridinyl)methanol
Supplier: Apollo Scientific
Warning
Synonyms: 6-Chloropyridine-3-methanol, 2-Chloro-5-hydroxymethylpyridine, 6-Chloro-3-(hydroxymethyl)pyridine
OR11307-100G OR11307-25G OR11307-5G
APOSOR11307-100GEA 136 EUR
APOSOR11307-100G APOSOR11307-25G APOSOR11307-5G
(6-Chloro-3-pyridinyl)methanol
(6-Chloro-3-pyridinyl)methanol
Formula: C₆H₆ClNO
MW: 143.57 g/mol
Boiling Pt: 120 °C/0.4mmHg
Melting Pt: 46…49 °C
Density: 1.28 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01075226
CAS Number: 21543-49-7