Ethyl methylsulphonylacetate 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Ethyl Methylsulphonylacetate, Ethyl 2-(methylsulphonyl)acetate
OR11368-25G
OR11368-5G
OR11368-100G
APOSOR11368-25GEA
92.8
EUR
APOSOR11368-25G
APOSOR11368-5G
APOSOR11368-100G
Ethyl methylsulphonylacetate 97%
Ethyl methylsulphonylacetate
Formula:
C₅H₁₀O₄S Boiling Pt: 111…113 °C/0.2mmHg Density: 1.234 g/cm³ (25 °C) Flash Pt: 109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041258 CAS Number: 4455-15-6 EINECS: 224-702-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...