4,4'-Dimethylbenzophenone
Supplier: Apollo Scientific
Warning
OR11371-100G OR11371-25G
APOSOR11371-100GEA 37.7 EUR
APOSOR11371-100G APOSOR11371-25G
4,4'-Dimethylbenzophenone
4,4'-Dimethylbenzophenone
Formula: C₁₅H₁₄O
MW: 210.28 g/mol
Boiling Pt: 200 °C/17mmHg
Melting Pt: 93…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00017214
CAS Number: 611-97-2
EINECS: 210-287-6