4-Amino-m-toluic acid 98%
Supplier: Apollo Scientific
Warning
OR11499-100G OR11499-25G
APOSOR11499-100GEA 60.1 EUR
APOSOR11499-100G APOSOR11499-25G
4-Amino-m-toluic acid 98%
4-Amino-m-toluic acid
Formula: C₈H₉NO₂
Melting Pt: 168…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00007736
CAS Number: 2486-70-6