6,7-Dichloroquinoxaline-2,3-dione 98%
Supplier: Apollo Scientific
Warning
Synonyms: 6,7-Dichloroquinoxaline-2,3(1H,4H)-dione, 6,7-Dichloro-1,4-dihydroquinoxaline-2,3-dione
OR11519-5G OR11519-25G
APOSOR11519-5GEA 65.4 EUR
APOSOR11519-5G APOSOR11519-25G
6,7-Dichloroquinoxaline-2,3-dione 98%
6,7-Dichloroquinoxaline-2,3-dione
Formula: C₈H₄Cl₂N₂O₂
MW: 231.0356 g/mol
Melting Pt: >310 °C
Storage Temperature: Ambient
MDL Number: MFCD00078575
CAS Number: 25983-13-5