5-Chlorosalicylaldehyde 97%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Chloro-2-hydroxybenzaldehyde
OR1167-100G OR1167-25G OR1167-500G
APOSOR1167-100GEA 48 EUR
APOSOR1167-100G APOSOR1167-25G APOSOR1167-500G
5-Chlorosalicylaldehyde 97%
5-Chlorosalicylaldehyde
Formula: C₇H₅ClO₂
Melting Pt: 99…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00003331
CAS Number: 635-93-8
EINECS: 211-244-4
UN: 3077
ADR: 9+EHS,III