3,5-Dibromoaniline 98%
Supplier: Apollo Scientific
Warning
OR11698-25G OR11698-5G OR11698-100G
APOSOR11698-25GEA 72.5 EUR
APOSOR11698-25G APOSOR11698-5G APOSOR11698-100G
3,5-Dibromoaniline 98%
3,5-Dibromoaniline
Formula: C₆H₅Br₂N
Melting Pt: 55…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00047841
CAS Number: 626-40-4